COMH93D5F4FA Combi-Blocks, Inc.

Methyltetrazine-peg12-dbco

  • MDL number MFCD31561138
  • Cas Number : 2183440-28-8
  • Molecular Weight : 1003.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1326
Availability details for
Quantity
  • Availability
  • Add to Cart