COMH93D5F509 Combi-Blocks, Inc.

(3-(N,N-Dimethylsulfamoyl)-4-methoxyphenyl)boronic acid

  • MDL number MFCD28400499
  • Cas Number : 874459-70-8
  • Molecular Weight : 259.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1346
Availability details for
Quantity
  • Availability
  • Add to Cart