COMH93D5F525 Combi-Blocks, Inc.

(R)-2,3-Dihydroxy-3-methyl-butyric acid benzyl ester

  • MDL number MFCD29086947
  • Cas Number : 184528-76-5
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1380
Availability details for
Quantity
  • Availability
  • Add to Cart