COMH93D5F531 Combi-Blocks, Inc.

(3-Iodo-phenyl)-acetic acid tert-butyl ester

  • MDL number MFCD30609617
  • Cas Number : 2206970-15-0
  • Molecular Weight : 318.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1397
Availability details for
Quantity
  • Availability
  • Add to Cart