COMH93D5F53A Combi-Blocks, Inc.

(2S)-1-(Methylamino)propan-2-ol hydrochloride

  • MDL number MFCD28397712
  • Cas Number : 1807937-56-9
  • Molecular Weight : 125.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1406

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1406
Availability details for
Quantity
  • Availability
  • Add to Cart