COMH93D5F549 Combi-Blocks, Inc.

3,6-Dibromo-9-(4-octoxyphenyl)carbazole

  • MDL number MFCD30536723
  • Cas Number : 917773-26-3
  • Molecular Weight : 529.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1422

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1422
Availability details for
Quantity
  • Availability
  • Add to Cart