COMH93D5F56E Combi-Blocks, Inc.

1-(3-Bromo-5-methylbenzoyl)pyrrolidine

  • MDL number MFCD27728741
  • Cas Number : 1873196-38-3
  • Molecular Weight : 268.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1468
Availability details for
Quantity
  • Availability
  • Add to Cart