COMH93D5F571 Combi-Blocks, Inc.

tert-Butyl (r)-(1-(3-bromopyridin-2-yl)ethyl)carbamate

  • MDL number MFCD30471324
  • Cas Number : 2007925-19-9
  • Molecular Weight : 301.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1471
Availability details for
Quantity
  • Availability
  • Add to Cart