COMH93D5F575 Combi-Blocks, Inc.

1-(4-Methoxy-benzyl)-3,5-dimethyl-1h-pyrazol-4-ol

  • MDL number MFCD31583220
  • Cas Number : 2270908-99-9
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1476
Availability details for
Quantity
  • Availability
  • Add to Cart