COMH93D5F584 Combi-Blocks, Inc.

tert-Butyl ((3-oxoisoindolin-5-yl)methyl)carbamate

  • MDL number MFCD28142682
  • Cas Number : 1312536-57-4
  • Molecular Weight : 262.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1495
Availability details for
Quantity
  • Availability
  • Add to Cart