COMH93D5F62C Combi-Blocks, Inc.

Ethyl 2,3,4,5-tetrahydro-1h-pyrrolo[1,2-a][1,4]diazepine-8-carboxylate hcl

  • MDL number MFCD29058599
  • Cas Number : 1338563-18-0
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1702

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1702
Availability details for
Quantity
  • Availability
  • Add to Cart