COMH93D5F672 Combi-Blocks, Inc.

Ethyl 2-amino-2-(10,11-dihydro-5H-dibenzo[a,d][7]annulen-5-yl)acetate HCl

  • MDL number MFCD30471993
  • Cas Number : 147900-32-1
  • Molecular Weight : 331.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1792
Availability details for
Quantity
  • Availability
  • Add to Cart