COMH93D5F693 Combi-Blocks, Inc.

Nvp-bsk805 2hcl

  • MDL number MFCD22420810
  • Cas Number : 1942919-79-0
  • Molecular Weight : 563.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1832
Availability details for
Quantity
  • Availability
  • Add to Cart