COMH93D5F6B5 Combi-Blocks, Inc.

(Meso-1r,5s,6s)-3-oxabicyclo[3.1.0]hexan-6-amine hcl

  • MDL number MFCD30828738
  • Molecular Weight : 135.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1874

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1874
Availability details for
Quantity
  • Availability
  • Add to Cart