COMH93D5F6C5 Combi-Blocks, Inc.

Methyl 1-(2-chlorophenyl)-5-(5-(3-(methylsulfonyl)phenyl)-1,3,4-thiadiazol-2-yl)-1h-pyrazole-3-carboxylate

  • MDL number MFCD30730108
  • Cas Number : 1454288-71-1
  • Molecular Weight : 474.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1891

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1891
Availability details for
Quantity
  • Availability
  • Add to Cart