COMH93D5F717 Combi-Blocks, Inc.

4-Phenylpyridin-3-amine hydrochloride

  • MDL number MFCD23099096
  • Cas Number : 1798733-74-0
  • Molecular Weight : 206.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1986
Availability details for
Quantity
  • Availability
  • Add to Cart