COMH93D5F739 Combi-Blocks, Inc.

Sodium 1-ethoxy-1,3-dioxopropan-2-ide

  • MDL number MFCD31555427
  • Cas Number : 2135339-48-7
  • Molecular Weight : 138.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2029
Availability details for
Quantity
  • Availability
  • Add to Cart