COMH93D5F747 Combi-Blocks, Inc.

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,6-bis(trifluoromethyl)pyridine

  • MDL number MFCD26516889
  • Cas Number : 1320747-32-7
  • Molecular Weight : 341.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2045
Availability details for
Quantity
  • Availability
  • Add to Cart