COMH93D5F74D Combi-Blocks, Inc.

2-Nitro-5-phenoxypyridine

  • MDL number MFCD23161847
  • Cas Number : 779345-38-9
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2051
Availability details for
Quantity
  • Availability
  • Add to Cart