COMH93D5F78D Combi-Blocks, Inc.

(R)-Benzyl 2-((tert-butoxycarbonyl)amino)-5-(dimethylamino)-5-oxopentanoate

  • MDL number MFCD30828778
  • Cas Number : 721927-49-7
  • Molecular Weight : 364.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2128

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2128
Availability details for
Quantity
  • Availability
  • Add to Cart