COMH93D5F7A2 Combi-Blocks, Inc.

1-(3,3-Difluoropiperidin-1-yl)-2-(piperidin-4-yl)ethan-1-one

  • MDL number MFCD29994851
  • Cas Number : 1894440-94-8
  • Molecular Weight : 246.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2151

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2151
Availability details for
Quantity
  • Availability
  • Add to Cart