COMH93D5F7C0 Combi-Blocks, Inc.

tert-Butyl-5-(hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate

  • MDL number MFCD30828941
  • Cas Number : 2089651-32-9
  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2190
Availability details for
Quantity
  • Availability
  • Add to Cart