COMH93D5F7D4 Combi-Blocks, Inc.

2-(2-Chloro-4-morpholinothieno[3,2-d]pyrimidin-6-yl)propan-2-ol

  • MDL number MFCD28966046
  • Cas Number : 1033743-25-7
  • Molecular Weight : 313.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2212

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2212
Availability details for
Quantity
  • Availability
  • Add to Cart