COMH93D5F7EA Combi-Blocks, Inc.

4-Methyl-2-phenylmorpholine

  • MDL number MFCD31556253
  • Cas Number : 21532-10-5
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2244

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2244
Availability details for
Quantity
  • Availability
  • Add to Cart