COMH93D5F7F4 Combi-Blocks, Inc.

5-Iodo-n-methoxy-n-methyl-1h-indazole-3-carboxamide

  • MDL number MFCD22689542
  • Cas Number : 351457-14-2
  • Molecular Weight : 331.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2254
Availability details for
Quantity
  • Availability
  • Add to Cart