COMH93D5F82A Combi-Blocks, Inc.

tert-Butyl 3-(tert-butoxycarbonyl(2-hydroxyethyl)amino)propanoate

  • MDL number MFCD30490567
  • Cas Number : 1629737-97-8
  • Molecular Weight : 289.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2323

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2323
Availability details for
Quantity
  • Availability
  • Add to Cart