COMH93D5F856 Combi-Blocks, Inc.

3-Chloro-6-(dichloromethyl)pyridazine

  • MDL number MFCD31651816
  • Cas Number : 2304584-07-2
  • Molecular Weight : 197.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2386

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2386
Availability details for
Quantity
  • Availability
  • Add to Cart