COMH93D5F8B2 Combi-Blocks, Inc.

3-(Dimethylamino)-2,2-difluoropropanoic acid

  • MDL number MFCD28656233
  • Cas Number : 1781638-68-3
  • Molecular Weight : 153.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2502
Availability details for
Quantity
  • Availability
  • Add to Cart