COMH93D5F8F3 Combi-Blocks, Inc.

Benzyl (3r,4r)-3-((tert-butoxycarbonyl)amino)-4-fluoropiperidine-1-carboxylate

  • MDL number MFCD28010927
  • Cas Number : 1052713-38-8
  • Molecular Weight : 352.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2587
Availability details for
Quantity
  • Availability
  • Add to Cart