COMH93D5F912 Combi-Blocks, Inc.

(3Ar,4r,6as)-tert-butyl 4-(benzyloxycarbonylamino)-3a-fluorohexahydrocyclopenta[c]pyrrole-2(1h)-carboxylate

  • MDL number MFCD22209656
  • Cas Number : 1391728-16-7
  • Molecular Weight : 378.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2630

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2630
Availability details for
Quantity
  • Availability
  • Add to Cart