COMH93D5F925 Combi-Blocks, Inc.

tert-Butyl (6-chloro-[1,2,4]triazolo[4,3-b]pyridazin-8-yl)carbamate

  • MDL number MFCD28041162
  • Cas Number : 1263425-79-1
  • Molecular Weight : 269.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2649
Availability details for
Quantity
  • Availability
  • Add to Cart