COMH93D5F941 Combi-Blocks, Inc.

1,1,1,3,3,3-Hexafluoropropan-2-yl piperazine-1-carboxylate

  • MDL number MFCD30803196
  • Cas Number : 1446758-21-9
  • Molecular Weight : 280.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2680
Availability details for
Quantity
  • Availability
  • Add to Cart