COMH93D5F958 Combi-Blocks, Inc.

3-[2-(tert-Butyl-dimethylsilanyloxy)ethoxy]propane-1,2-diol

  • MDL number MFCD30486303
  • Cas Number : 2168359-06-4
  • Molecular Weight : 250.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2708

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2708
Availability details for
Quantity
  • Availability
  • Add to Cart