COMH93D5F971 Combi-Blocks, Inc.

2-Bromo-1-(4-chlorophenyl)-2-cyclopropylethanone

  • MDL number MFCD30472050
  • Cas Number : 2007908-49-6
  • Molecular Weight : 273.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2739
Availability details for
Quantity
  • Availability
  • Add to Cart