COMH93D5F98D Combi-Blocks, Inc.

6,6-Difluoro-1,4-diazepane dihydrochloride

  • MDL number MFCD28902474
  • Cas Number : 1956307-23-5
  • Molecular Weight : 209.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-2772
Availability details for
Quantity
  • Availability
  • Add to Cart