COMH93D5FA85 Combi-Blocks, Inc.

(3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

  • MDL number MFCD31556773
  • Cas Number : 1857323-39-7
  • Molecular Weight : 270.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3072
Availability details for
Quantity
  • Availability
  • Add to Cart