COMH93D5FAAE Combi-Blocks, Inc.

5-(tert-Butyl)pyrimidin-2-amine hcl

  • MDL number MFCD30377224
  • Cas Number : 1958100-57-6
  • Molecular Weight : 187.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3120

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3120
Availability details for
Quantity
  • Availability
  • Add to Cart