COMH93D5FAB3 Combi-Blocks, Inc.

5-Chloro-3-methoxy-2-nitropyridine

  • MDL number MFCD22418344
  • Cas Number : 152684-28-1
  • Molecular Weight : 188.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3127

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3127
Availability details for
Quantity
  • Availability
  • Add to Cart