COMH93D5FAC4 Combi-Blocks, Inc.

4,6-Dichloroisophthalonitrile

  • MDL number MFCD30481248
  • Cas Number : 1897-44-5
  • Molecular Weight : 197.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3147
Availability details for
Quantity
  • Availability
  • Add to Cart