COMH93D5FAE5 Combi-Blocks, Inc.

tert-Butyl (1r,3r,5s)-3-(methylamino)-9-azabicyclo[3.3.1]nonane-9-carboxylate

  • MDL number MFCD30471680
  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3186

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3186
Availability details for
Quantity
  • Availability
  • Add to Cart