COMH93D5FB64 Combi-Blocks, Inc.

(R)-(2-Chloro-3-(trifluoromethyl)phenyl)(1-(5-fluoropyrimidin-2-yl)-4-methyl-1,4,6,7-tetrahydro-5h-[1,2,3]triazolo[4,5-c]pyridin-5-yl)methanone

  • MDL number MFCD31556312
  • Cas Number : 1627902-21-9
  • Molecular Weight : 440.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3342

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3342
Availability details for
Quantity
  • Availability
  • Add to Cart