COMH93D5FBB0 Combi-Blocks, Inc.

3-(4-((1H-Imidazol-1-yl)methyl)phenyl)-n-(tert-butyl)thiophene-2-sulfonamide

  • MDL number MFCD30728487
  • Cas Number : 2089650-46-2
  • Molecular Weight : 375.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3431
Availability details for
Quantity
  • Availability
  • Add to Cart