COMH93D5FBD7 Combi-Blocks, Inc.

[(4-Bromo-3-fluorophenyl)methyl][(2-fluorophenyl)methyl]amine

  • MDL number MFCD25567833
  • Cas Number : 1480180-18-4
  • Molecular Weight : 312.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3483
Availability details for
Quantity
  • Availability
  • Add to Cart