COMH93D5FBEA Combi-Blocks, Inc.

Desmethyl doxylamine

  • MDL number MFCD31562535
  • Cas Number : 1221-70-1
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3508

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3508
Availability details for
Quantity
  • Availability
  • Add to Cart