COMH93D5FC04 Combi-Blocks, Inc.

2-Bromo-4-(2,2-difluoroethoxy)-1-methylbenzene

  • MDL number MFCD30263451
  • Cas Number : 1861177-37-8
  • Molecular Weight : 251.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3541
Availability details for
Quantity
  • Availability
  • Add to Cart