COMH93D5FC24 Combi-Blocks, Inc.

Methyl 3-methoxyquinoline-6-carboxylate

  • MDL number MFCD28141138
  • Cas Number : 1051316-27-8
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3577
Availability details for
Quantity
  • Availability
  • Add to Cart