COMH93D5FC28 Combi-Blocks, Inc.

4-Methyl-2-oxo-1,2,3,4-tetrahydroquinoxaline-6-boronic acid pinacol ester

  • MDL number MFCD31001395
  • Molecular Weight : 288.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3582

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3582
Availability details for
Quantity
  • Availability
  • Add to Cart