COMH93D5FC4B Combi-Blocks, Inc.

(4-(1-(4-Methylpiperazin-1-yl)ethyl)phenyl)boronic acid

  • MDL number MFCD28400320
  • Cas Number : 1704069-65-7
  • Molecular Weight : 248.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3628

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3628
Availability details for
Quantity
  • Availability
  • Add to Cart