COMH93D5FCDB Combi-Blocks, Inc.

6-(Trifluoromethoxy)pyridin-3-ol

  • MDL number MFCD25450574
  • Cas Number : 1361494-43-0
  • Molecular Weight : 179.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3801
Availability details for
Quantity
  • Availability
  • Add to Cart