COMH93D5FD06 Combi-Blocks, Inc.

4-[[4-(4-Morpholinyl)butyl]thio]-phenol hcl

  • MDL number MFCD22543912
  • Cas Number : 57055-96-6
  • Molecular Weight : 303.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3854
Availability details for
Quantity
  • Availability
  • Add to Cart